Methyl 4-chloroacetoacetate, 97%
Methyl 4-chloroacetoacetate, 97%
Methyl 4-chloroacetoacetate, 97%
Thermo Scientific Chemicals

Methyl 4-chloroacetoacetate, 97%

CAS: 32807-28-6 | C5H7ClO3 | 150.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number 241621000
Price (USD)
-
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS32807-28-6
IUPAC Namemethyl 4-chloro-3-oxobutanoate
Molecular FormulaC5H7ClO3
InChI KeyHFLMYYLFSNEOOT-UHFFFAOYSA-N
SMILESCOC(=O)CC(=O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=96.0 %
Refractive index1.4560 to 1.4580 (20°C, 589 nm)
Appearance (Color)Clear light yellow to light orange
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only