Silica gel, for chromatography, 0.035-0.070 mm, 90 A
Silica gel, for chromatography, 0.035-0.070 mm, 90 A
Silica gel, for chromatography, 0.035-0.070 mm, 90 A
Thermo Scientific Chemicals

Silica gel, for chromatography, 0.035-0.070 mm, 90 A

CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic Bottle
Catalog number 241652500
Price (USD)
42.65
Special offer
Online exclusive
Ends: 30-Jun-2026
49.90
Save 7.25 (15%)
Each
Quantity:
250 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
42.65
Special offer
Online exclusive
Ends: 30-Jun-2026
49.90
Save 7.25 (15%)
Each
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
pH6.5 to 7.5 (5 % w/w suspension)
Particle size35 to 70 µm
Appearance (Form)Powder
Bulk density385 to 440 g/l
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatography
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only