2-Hydroxy-6-aminopurine, 98%
2-Hydroxy-6-aminopurine, 98%
2-Hydroxy-6-aminopurine, 98%
Acros Organics

2-Hydroxy-6-aminopurine, 98%

CAS: 3373-53-3 | C5H5N5O | 151.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
241752500
also known as 24175
250 mgGlass Bottle
Catalog number 241752500
also known as 24175
HSN 29335990
Price (INR)
16,008.00
Each
Quantity:
250 mg
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
16,008.00
Each
Chemical Identifiers
CAS3373-53-3
IUPAC Name6-amino-2,7-dihydro-1H-purin-2-one
Molecular FormulaC5H5N5O
InChI KeyDRAVOWXCEBXPTN-UHFFFAOYSA-N
SMILESNC1=C2NC=NC2=NC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light beige
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only