2,3-Dichloro-5-(trifluoromethyl)pyridine, 97%
2,3-Dichloro-5-(trifluoromethyl)pyridine, 97%
2,3-Dichloro-5-(trifluoromethyl)pyridine, 97%
Thermo Scientific Chemicals

2,3-Dichloro-5-(trifluoromethyl)pyridine, 97%

CAS: 69045-84-7 | C6H2Cl2F3N | 215.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 241811000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS69045-84-7
IUPAC Name2,3-dichloro-5-(trifluoromethyl)pyridine
Molecular FormulaC6H2Cl2F3N
InChI KeyABNQGNFVSFKJGI-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CN=C(Cl)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.4745 to 1.4765 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only