Product Image
Thermo Scientific Chemicals

Pimozide

 
Have Questions?
Catalog NumberQuantity
2420100101 g
Catalog number 242010010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS2062-78-4
IUPAC Name1-{1-[4,4-bis(4-fluorophenyl)butyl]piperidin-4-yl}-2,3-dihydro-1H-1,3-benzodiazol-2-one
Molecular FormulaC28H29F2N3O
InChI KeyYVUQSNJEYSNKRX-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1)C(CCCN1CCC(CC1)N1C(=O)NC2=CC=CC=C12)C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
Melting point214°C to 222°C
Appearance (Color)White to light yellow
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only