Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles
Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles
Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles
Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles
Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles
Thermo Scientific™

Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles

Meet regulatory labeling requirements for chemical containers. These Right-to-Know Wash Bottles use standard, instantly recognizable symbols and colors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Chemical LabelUnit Size
2425-0501500 mLAcetoneCase of 24
2425-0502500 mLEthyl AlcoholCase of 24
2425-0502PK
also known as 2425-0502
500 mLEthyl AlcoholPack of 6
2425-0503500 mLMethanolCase of 24
2425-0503PK
also known as 2425-0503
500 mLMethanolPack of 6
2425-0506500 mLSodium HypochloriteCase of 24
2425-0506PK
also known as 2425-0506
500 mLSodium HypochloritePack of 6
2425-0504500 mLIsopropanolCase of 24
2425-0504PK
also known as 2425-0504
500 mLIsopropanolPack of 6
2425-0505500 mLDistilled WaterCase of 24
2425-0505PK
also known as 2425-0505
500 mLDistilled WaterPack of 6
2425-0500500 mLAssortedCase of 20
2425-0500PK
also known as 2425-0500
500 mLAssortedPack of 5
2425-0501PK
also known as 2425-0501
500 mLAcetonePack of 6
2425-10021000 mLEthyl AlcoholCase of 12
2425-1002PK
also known as 2425-1002
1000 mLEthyl AlcoholPack of 4
2425-10031000 mLMethanolCase of 12
2425-1003PK1000 mLMethanolPack of 4
2425-10041000 mLIsopropanolCase of 12
2425-1004PK
also known as 2425-1004
1000 mLIsopropanolPack of 4
2425-10051000 mLDistilled WaterCase of 12
2425-1005PK
also known as 2425-1005
1000 mLDistilled WaterPack of 4
Catalog number 2425-0501
Price (USD)
344.70
Online exclusive
402.00
Save 57.30
Case of 24
Capacity (Metric):
500 mL
Chemical Label:
Acetone
Unit Size:
Case of 24
Price (USD)
344.70
Online exclusive
402.00
Save 57.30
Case of 24

Meet NFPA and ICS labeling standards for chemical containers with Thermo Scientific™ Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles, which use standard, instantly recognizable colors and GHS symbols. These low-density polyethylene bottles are flexible and easy to squeeze for ergonomic dispensing. Made from high quality laboratory grade plastic materials for dependably low leachables and extractables.

Features:

  • Polypropylene or high-density polyethylene closure; polypropylene copolymer fill tube
  • Self-venting, so contents stay in bottle; prevents self-dispensing
  • Chemical name, International Chemical Society (ICS) chemical formula and Chemical Abstract Service (CAS) number printed on bottle for easy identification
  • Color bar that corresponds to colored closure to prevent cross-contamination
  • Designed to dispense through the top with a closure and stem molded in one piece for uniform service
  • Tip that can be cut back to increase flow
  • Tight fit of draw tube and closure that prevents separation when squeezing bottle

Ordering Information:

Available in packs of the same 16 or 32 oz. (500 or 1000mL) bottle, or in a five-bottle (16 oz./500mL only) assortment pack consisting of one each acetone, ethyl alcohol, isopropanol, methanol and distilled water.

.

Specifications
AutoclavableNot Autoclavable
Capacity16 oz., 500 mL
Capacity (English)16 oz.
Certifications/ComplianceMeet requirements of OSHA Hazard Communication Regulation [29 CFR 1910.1200(f)(4)].
Closure ColorRed
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
DescriptionRTK Wash Bottle
Diameter (Metric)72.64 mm
Height (English)8.3 in.
Height (Metric)210.82 mm
No. per Case24
No. per Pack6
Product TypeSafety Wash Bottle
ShapeCylindrical
Through the Closure DispensingYes
Capacity (Metric)500 mL
Chemical LabelAcetone
Closure IncludedYes
ColorNatural
GraduatedNo
MaterialLDPE
MouthNarrow
Unit SizeCase of 24