384 Well Plate, Non-Treated Surface, Pack of 1 - FAQs
384 Well Plate, Non-Treated Surface, Pack of 1 - FAQs
View additional product information for Nunc™ 384-Well, Non-Treated, Flat-Bottom Microplate - FAQs (242765, 265202, 242757)
1 product FAQs found
Should I use an opaque black/white immuno plate, or an "optical-bottom" design?
Opaque plates can only be used on “top reader” instruments, which detect (or, in fluorescence mode, excite and detect) from above the plate. Readers that detect from underneath the plate (or imaging systems) use optical-bottom plates, which have a black or white upper well structure and a transparent (polystyrene film or optical coverglass) bottom.