N-(9H-Fluoren-2-ylmethoxycarbonyloxy)succinimide, 98%
N-(9H-Fluoren-2-ylmethoxycarbonyloxy)succinimide, 98%
N-(9H-Fluoren-2-ylmethoxycarbonyloxy)succinimide, 98%
Thermo Scientific Chemicals

N-(9H-Fluoren-2-ylmethoxycarbonyloxy)succinimide, 98%

CAS: 82911-69-1 | C19H15NO5 | 337.29 g/mol
Have Questions?
Catalog NumberQuantityPackaging
2431000505 gGlass bottle
Catalog number 243100050
Price (USD)
73.65
Special offer
Online exclusive
Ends: 28-Dec-2026
86.60
Save 12.95
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
73.65
Special offer
Online exclusive
Ends: 28-Dec-2026
86.60
Save 12.95
Each
Chemical Identifiers
CAS82911-69-1
IUPAC Name2,5-dioxopyrrolidin-1-yl (9H-fluoren-9-yl)methyl carbonate
Molecular FormulaC19H15NO5
InChI KeyWMSUFWLPZLCIHP-UHFFFAOYSA-N
SMILESO=C(OCC1C2=CC=CC=C2C2=CC=CC=C12)ON1C(=O)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to off-white
HPLC>=97.5 %
Appearance (Form)Powder
Melting point145°C to 153°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only