3-Butyn-2-ol, 97%
3-Butyn-2-ol, 97%
3-Butyn-2-ol, 97%
Thermo Scientific Chemicals

3-Butyn-2-ol, 97%

CAS: 2028-63-9 | C4H6O | 70.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 245310100
Price (USD)
39.65
Special offer
Online exclusive
Ends: 28-Dec-2026
46.70
Save 7.05
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
39.65
Special offer
Online exclusive
Ends: 28-Dec-2026
46.70
Save 7.05
Each
Chemical Identifiers
CAS2028-63-9
IUPAC Namebut-3-yn-2-ol
Molecular FormulaC4H6O
InChI KeyGKPOMITUDGXOSB-UHFFFAOYNA-N
SMILESCC(O)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.4250 to 1.4270 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only