2',4'-Difluoroacetophenone, 99%
2',4'-Difluoroacetophenone, 99%
2',4'-Difluoroacetophenone, 99%
Thermo Scientific Chemicals

2',4'-Difluoroacetophenone, 99%

CAS: 364-83-0 | C8H6F2O | 156.12 g/mol
Have Questions?
Catalog NumberQuantity
24677010010 g
Catalog number 246770100
Price (USD)
110.00
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
110.00
Each
Chemical Identifiers
CAS364-83-0
IUPAC Name1-(2,4-difluorophenyl)ethan-1-one
Molecular FormulaC8H6F2O
InChI KeyQEWHNJPLPZOEKU-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(F)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay darken on storage
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4870 to 1.4890 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only