UltraPure™ 5M NaCl - FAQs
UltraPure™ 5M NaCl - FAQs
View additional product information for UltraPure™ 5M NaCl - FAQs (24740011)
1 product FAQs found
What can I do to prevent my DNA standards from smearing during electrophoresis?
Add 10 to 20 mM NaCl to the sample to maintain proper ionic strength.