Nunc™ NucleoLink™ Strips - FAQs
View additional product information for Nunc™ NucleoLink™ Strips - FAQs (248259)
2 product FAQs found
What is the shelf life of your Nunc NucleoLink Strips if I store them in a sealed ethylene bag at 4 degrees C?
Uncoated Nunc NucleoLink Strips are stable for at least three years, if the original packing is intact. After "coating" the strips with a nucleic acid probe, we generally store the strips in a sealed ethylene bag at 4 degrees C. The stability may vary with the probe composition. Nunc NucleoLink Strips that are stored in a sealed bag at 4 degrees C are stable for at least 10 months.
What detection methods should I use when performing an in situ hybridization using Nunc NucleoLink Strips?
When performing an in situ hybridization using Nunc NucleoLink Strips, we recommend the following detection methods:
- Fluorometric
- Colorimetric
- Radioactive probe