Nunc™ 96-Well Polystyrene Conical Bottom MicroWell™ Plates
Thermo Scientific™
Nunc™ 96-Well Polystyrene Conical Bottom MicroWell™ Plates
Recover samples completely with these conical well bottom plates. The bottom shape allows for easy pipette access.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityLidNo. per Case
249662SterileWithout Lid50/Cs.
249570Non-sterileWithout Lid180/Cs.
249935SterileWith Lid80/Cs.
249940SterileWithout Lid80/Cs.
249952SterileWith Lid160/Cs.
Catalog number 249662
Price (USD)
247.65
Online exclusive
272.00
Save 24.35
Case of 50
Sterility:
Sterile
Lid:
Without Lid
No. per Case:
50/Cs.
Price (USD)
247.65
Online exclusive
272.00
Save 24.35
Case of 50
Recover samples completely with Thermo Scientific™ Nunc™ 96-Well Polystyrene Conical Bottom MicroWell™ Plates. The bottom shape allows for easy pipette access. The plates are optimized for robotics, automated readers and liquid handling systems.
Features:
  • Conical bottom (V) microplates are ideal for sample recovery.
    • Applications include antibiotic screens, serological tests, storing and screening compounds, DNA libraries, and titration of solutions.
Warranty: 90 days
WARNING: Cancer – www.P65Warnings.ca.gov
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
FormatStandard
LidWithout Lid
No. per Case50/Cs.
No. per Pack1 Ea.
Packaging50 Cases
Product TypeMicroplate
Quantity1 Ea.
Skirt StyleLow Flange Pinchbar
Volume (Metric) Well300 μL
Array8 x 12
BarcodeNo Barcode
ColorClear
MaterialPolystyrene
No. of Wells96
SterilitySterile
Surface TreatmentNon-treated
Well ShapeConical Bottom (V-well)
Unit SizeCase of 50