3-Iodophenol, 99%
3-Iodophenol, 99%
3-Iodophenol, 99%
Thermo Scientific Chemicals

3-Iodophenol, 99%

CAS: 626-02-8 | C6H5IO | 220 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 249700050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS626-02-8
IUPAC Name3-iodophenol
Molecular FormulaC6H5IO
InChI KeyFXTKWBZFNQHAAO-UHFFFAOYSA-N
SMILESOC1=CC=CC(I)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)or fused solid
Melting point37°C to 43°C
Appearance (Color)White to yellow to brown or grey or purple
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only