Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Thermo Scientific™
Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Green features
Store samples safely with these low-binding 96-well polypropylene plates, which will not allow DNA or proteins to bind and will allow for complete sample recovery.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSterilityWell ShapeFit for Purpose
249950BlueNon-sterileRound, Conical Bottom (V-well)Certified Quality
249943RedNon-sterileRound, Conical Bottom (V-well)Certified Quality
249944NaturalNon-sterileRound, Conical Bottom (V-well)Certified Quality
249945BlackNon-sterileRound, Conical Bottom (V-well)Certified Quality
249946NaturalSterileRound, Conical Bottom (V-well)Sterile
249947YellowNon-sterileRound, Conical Bottom (V-well)Certified Quality
249949WhiteNon-sterileRound, Conical Bottom (V-well)Certified Quality
267245NaturalNon-sterileRoundCertified Quality
267334NaturalSterileRoundCertified Quality
267342BlackNon-sterileRoundCertified Quality
267350WhiteNon-sterileRoundCertified Quality
267385BlueNon-sterileRoundCertified Quality
267407YellowNon-sterileRoundCertified Quality
442587NaturalNon-sterileRound, Conical Bottom (V-well)Certified Quality
Catalog number 249950
Price (USD)
584.08
Online exclusive
721.67
Save 137.59
Case of 120
Color:
Blue
Sterility:
Non-sterile
Well Shape:
Round, Conical Bottom (V-well)
Fit for Purpose:
Certified Quality
Price (USD)
584.08
Online exclusive
721.67
Save 137.59
Case of 120
Achieve complete sample recovery with Thermo ScientificTM NuncTM 96-Well Polypropylene MicroWellTM Plates, which are the ideal storage plate as polypropylene has a lower binding capacity so proteins or DNA will not bind. Withstands temperatures from -80° to +121°C. Available in a variety of colors for quick identification of storage plates.
Features:
  • Shared wall design increases the well volume
  • Wider wells and rounded bottom improve mixing
  • Conical bottom well for optimal sample recovery
  • Black and white plates available for fluorescent- and luminescent-based applications
  • Low binding polypropylene for homogeneous assays and storage
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
AutoclavableAutoclavable
Binding TypeLow binding PP
Certifications/ComplianceCertified RNase and DNase Free. ANSI Compliant.
Dnase FreeYes
Description96-well Polypropylene Sample Processing & Storage Plates
For Use With (Application)Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
For Use With (Equipment)ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
FormatStandard
LidWithout Lid
No. per Case120
No. per Pack20
Packaging20 pk., 120 cs.
Plate SizeANSI/ SLAS Standard
Product Type96-well plate
Quantity120/Cs.
RNase FreeYes
Regulatory StatusRUO
Volume (Metric) Well450 μL
Volume (Metric) Working400 μL
Array8 x 12
BarcodeCustom Barcoding Available
ColorBlue
Fit for PurposeCertified Quality
MaterialPolypropylene
No. of Wells96
Product LineNunc
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeRound, Conical Bottom (V-well)
Unit SizeCase of 120