eBioscience™ Streptavidin PE-Cyanine7 Conjugate
Invitrogen™

eBioscience™ Streptavidin PE-Cyanine7 Conjugate

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.
Have Questions?
Catalog NumberQuantity
25-4317-82100 μg
Catalog number 25-4317-82
Price (USD)
243.65
Online exclusive
324.00
Save 80.35
Each
Quantity:
100 μg
Price (USD)
243.65
Online exclusive
324.00
Save 80.35
Each

The streptavidin fluorochrome conjugates are commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.

Laser

Blue Laser, Green Laser, Yellow-Green Laser

Emit

775 nm

Excite

488 - 561 nm

Reported Application

Flow Cytometric Analysis

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration0.2 mg/mL
Label or DyePE-Cy7
Product TypeStreptavidin Conjugate (Fluorescent)
Quantity100 μg
Target MoleculeBiotin
ConjugatePE-Cy7
FormLiquid
Product LineeBioscience
Unit SizeEach