Nunc™ Microplate Lids
Nunc™ Microplate Lids
Nunc™ Microplate Lids
Nunc™ Microplate Lids
Thermo Scientific™

Nunc™ Microplate Lids

Protect samples from contamination and evaporation during assay processing, incubation or storage with these microplate lids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPlate CompatibilitySterilityCondensation RingsNo. per Pack
250003All MicroplatesNon-sterileNo20
250002All MicroplatesSterileNo20
250005All MicroplatesNon-sterileNo20
253623All MicroplatesNon-sterileYes25
253624All MicroplatesSterileYes25
26333996-well PlateNon-sterileYes5
26412296-well PlateSterileYes1
264611384-well PlateSterileNo1
264612384-well PlateNon-sterileNo20
264616384-well PlateSterileNo20
26462396-well PlateSterileYes20
Catalog number 250003
Price (USD)
87.13
Online exclusive
174.25
Save 87.12
Case of 60
Plate Compatibility:
All Microplates
Sterility:
Non-sterile
Condensation Rings:
No
No. per Pack:
20
Price (USD)
87.13
Online exclusive
174.25
Save 87.12
Case of 60
Protect samples from contamination and evaporation during assay processing, incubation or storage with Thermo ScientificTM NuncTM Microplate Lids.
  • Lids are for use with any standard plates
  • 96 Well Lids have individual condensation rings
  • Lids have notches for quick orientation
  • Universal Lids can be used on any standard height plate on the market
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Condensation RingsNo
Cut-off CornersNo
Dimensions (L x W)5 x 3.35 in. (127 x 85 mm)
Dimensions (L x W x H)5 x 3.35 x 0.33 in. (127 x 85 x 8.34 mm)
Evaporation BarrierNo
Height (English)0.33 in.
Height (Metric)8.34 mm
Length (English)5 in.
Length (Metric)127 mm
No. per Case60
No. per Pack20
Product TypeUniversal Lid
Temperature Range (Metric)-20°C to +80°C
Well ShapeAll well shapes
Width (English)3.35 in.
Width (Metric)85 mm
AdhesiveNone
ColorClear
MaterialPolystyrene
No. of Wells384/1536
Plate CompatibilityAll Microplates
Product LineNunc
SterilityNon-sterile
Unit SizeCase of 60