n-Butyl lactate, 99%
n-Butyl lactate, 99%
n-Butyl lactate, 99%
Thermo Scientific Chemicals

n-Butyl lactate, 99%

CAS: 138-22-7 | C7H14O3 | 146.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 250180025
Price (USD)
341.65
Online exclusive
379.00
Save 37.35
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
341.65
Online exclusive
379.00
Save 37.35
Each
Chemical Identifiers
CAS138-22-7
IUPAC Namebutyl 2-hydroxypropanoate
Molecular FormulaC7H14O3
InChI KeyMRABAEUHTLLEML-UHFFFAOYNA-N
SMILESCCCCOC(=O)C(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Infrared spectrumConforms
Refractive index1.4200 to 1.4220 (20°C, 589 nm)
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only