Citric acid, disodium salt sesquihydrate, 99%, pure
Citric acid, disodium salt sesquihydrate, 99%, pure
Citric acid, disodium salt sesquihydrate, 99%, pure
Thermo Scientific Chemicals

Citric acid, disodium salt sesquihydrate, 99%, pure

CAS: 1545832 | C12H18Na4O17 | 526.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 250240010
Price (USD)
102.65
Special offer
Online exclusive
Ends: 28-Dec-2026
120.00
Save 17.35
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
102.65
Special offer
Online exclusive
Ends: 28-Dec-2026
120.00
Save 17.35
Each
Chemical Identifiers
CAS6132-05-4
IUPAC Nametetrasodium bis(2-(carboxymethyl)-2-hydroxybutanedioate) trihydrate
Molecular FormulaC12H18Na4O17
InChI KeyHGPVLOQNBSHYEI-UHFFFAOYNA-J
SMILESO.O.O.[Na+].[Na+].[Na+].[Na+].OC(=O)CC(O)(CC([O-])=O)C([O-])=O.OC(=O)CC(O)(CC([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Titration with HClO498.5 to 104.0 %
pH4.9 to 5.2 (3 % solution)
Appearance (Color)White
Insoluble matter=<0.01 % (in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only