Theophylline, 99+%, anhydrous
Theophylline, 99+%, anhydrous
Theophylline, 99+%, anhydrous
Thermo Scientific Chemicals

Theophylline, 99+%, anhydrous

CAS: 58-55-9 | C7H8N4O2 | 180.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number 250312500
Price (USD)
124.00
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
124.00
Each
Chemical Identifiers
CAS58-55-9
IUPAC Name1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular FormulaC7H8N4O2
InChI KeyZFXYFBGIUFBOJW-UHFFFAOYSA-N
SMILESCN1C2=C(NC=N2)C(=O)N(C)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting point270°C to 274°C
Infrared spectrumConforms
Titration with NaOH>=99.0 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only