Thermo Scientific Pierce Pyridine (PY) is a sequencing-grade preparation with high purity and consistent absorptivity properties for use in HPLCRead more
Have Questions?
Catalog Number
Quantity
25104
100 g
Catalog number 25104
Price (EUR)
112,84
Online exclusive
124,00
Save 11,16 (9%)
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
112,84
Online exclusive
124,00
Save 11,16 (9%)
Each
Thermo Scientific Pierce Pyridine (PY) is a sequencing-grade preparation with high purity and consistent absorptivity properties for use in HPLC and mass spectrometry methods.
Features of Pyridine (PY) :
• Water-miscible organic solvent for a variety of molecular and protein biology methods • High purity, low water, and no peroxides or aldehydes • Ninhydrin test ensures low amines • UV-absorption test confirms suitability for HPLC detection • Liquid reagent packaged by mass in amber glass bottles
Pierce pyridine is specially purified and each lot is tested to the highest specifications to ensure the integrity of your data, maximize sensitivity in your assay and to prolong the life of your equipment.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity100 g
For Use WithHPLC, Mass Spectrometry
FormLiquid
GradeLC-MS
Item NamePyridine
Product LinePierce
Reagent TypeSolvent (Pure)
Unit SizeEach
Contents & Storage
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -20°C.