Nunc™ Disposable Loops and Needles
Nunc™ Disposable Loops and Needles
Nunc™ Disposable Loops and Needles
Nunc™ Disposable Loops and Needles
Nunc™ Disposable Loops and Needles
Thermo Scientific™
Nunc™ Disposable Loops and Needles
Easily identify these color-coded disposable loops and needles, precision molded from nontoxic (USP Class VI) polystyrene material.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolumeQuantityUnit Size
25158610 μLCarton of 1000Case of 4000
251586PK
also known as 251586
10 μLCarton of 1000Pack of 1000
2532871 μLCarton of 1000Case of 4000
253287PK
also known as 253287
1 μLCarton of 1000Pack of 1000
2544101 μLCarton of 600Case of 2400
254410PK
also known as 254410
1 μLCarton of 600Pack of 600
25443710 μLCarton of 600Case of 2400
254437PK10 μLCarton of 600Pack of 600
Catalog number 251586
Price (USD)
880.99
Online exclusive
1,032.26
Save 151.27
Case of 4000
Volume:
10 μL
Quantity:
Carton of 1000
Unit Size:
Case of 4000
Price (USD)
880.99
Online exclusive
1,032.26
Save 151.27
Case of 4000
Easily identify color-coded Thermo Scientific™ Nunc™ Disposable Loops and Needles, precision molded from nontoxic (USP Class VI) polystyrene material to provide a smooth loop surface. Thermo Scientific Nunc products are high quality plastics for use in biotechnology, pharmaceutical and research laboratories, as well as in the production of vaccines and diagnostic kits.
Features:
  • Radiation sterilized—no need for flame sterilization
  • Hydrophilic treatment during manufacturing process
  • Liquid adherence properties that prevent accidental discharge of droplets
  • Loops available in 10 and 1 μL sizes
  • Meet semi-quantitative standard for handling samples
  • Calibration documents available for any batch upon request
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
ColorBlue
DisposableSingle-use
QuantityCarton of 1000
MaterialPolystyrene
Product TypeInoculating Loop (Blue)
SterilitySterile
Volume10 μL
Unit SizeCase of 4000