4-Amino-6-chloro-1,3-benzenedisulfonamide, 98%
4-Amino-6-chloro-1,3-benzenedisulfonamide, 98%
4-Amino-6-chloro-1,3-benzenedisulfonamide, 98%
Thermo Scientific Chemicals

4-Amino-6-chloro-1,3-benzenedisulfonamide, 98%

CAS: 121-30-2 | C6H8ClN3O4S2 | 285.73 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Packaging:
Glass bottle
Catalog number 251850050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS121-30-2
IUPAC Name4-amino-6-chlorobenzene-1,3-disulfonamide
Molecular FormulaC6H8ClN3O4S2
InChI KeyIHJCXVZDYSXXFT-UHFFFAOYSA-N
SMILESNC1=CC(Cl)=C(C=C1S(N)(=O)=O)S(N)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=97.5 %
Appearance (Color)White to off-white
Melting point260°C to 264°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only