Samco™ Narrow Stem Transfer Pipettes
Samco™ Narrow Stem Transfer Pipettes
Samco™ Narrow Stem Transfer Pipettes
Samco™ Narrow Stem Transfer Pipettes
Thermo Scientific™

Samco™ Narrow Stem Transfer Pipettes

Ideal for work with narrow tubes and containers with small openings.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityBulb Draw (Metric)Packaging TypeUnit Size
25215.3 mLNon-sterile11.6 mLBulkCase of 2500
2411.3 mLNon-sterile0.8 mLBulkCase of 4000
241PK
also known as 241
1.3 mLNon-sterile0.8 mLBulkPack of 500
241-20S1.3 mLSterile0.8 mL20/Bag, 500/Pk., 4000/Cs.Each
2424.3 mLNon-sterile3.1 mLBulkCase of 5000
242PK
also known as 242
4.3 mLNon-sterile3.1 mLBulkPack of 500
242-1S4.3 mLSterile3.1 mLIndividually WrappedCase of 4000
242-1SPK
also known as 242-1S
4.3 mLSterile3.1 mLIndividually WrappedPack of 500
242-20S4.3 mLSterile3.1 mL20/Bag, 760/Pk., 6080/Cs.Case of 6080
2514.5 mLNon-sterile3.3 mLBulkCase of 5000
251PK
also known as 251
4.5 mLNon-sterile3.3 mLBulkPack of 500
251-1S4.5 mLSterile3.3 mLIndividually WrappedCase of 4000
251-1SPK
also known as 251-1S
4.5 mLSterile3.3 mLIndividually WrappedPack of 500
251-20S4.5 mLSterile3.3 mL20/Bag, 500/Pk., 4000/Cs.Case of 500
252PK15.3 mLNon-sterile11.6 mLBulkPack of 250
252-1S15.3 mLSterile11.6 mLIndividually WrappedCase of 2000
252-1SPK
also known as 252-1S
15.3 mLSterile11.6 mLIndividually WrappedPack of 250
252-20S15.3 mLSterile11.6 mL20/Bag, 200/Pk., 2000/Cs.Case of 1600
694TS
also known as 694
6.9 mLNon-sterile3.2 mLBulkCase of 5000
694TSPK
also known as 694
6.9 mLNon-sterile3.2 mLBulkPack of 500
694-1S6.9 mLSterile3.2 mLIndividually WrappedEach
241-1S1.3 mLSterile0.8 mLIndividually WrappedEach
Catalog number 252
Price (USD)
445.50
Online exclusive
522.00
Save 76.50
Case of 2500
Volume (Metric):
15.3 mL
Sterility:
Non-sterile
Bulb Draw (Metric):
11.6 mL
Packaging Type:
Bulk
Unit Size:
Case of 2500
Price (USD)
445.50
Online exclusive
522.00
Save 76.50
Case of 2500
Eliminate plugging, packaging and autoclaving with Thermo Scientific™ Samco™ Narrow Stem Transfer Pipettes. These polyethylene pipettes prevent cross-contamination and are used narrow tubes and containers with small openings . The single-unit design features a built-in pipette bulb, and a flexible stem bends to sample from narrow or small tubes.
  • Made of shatterproof, nontoxic polyethylene
  • Low-affinity surface prevents proteins binding in research applications
  • Can be frozen in liquid nitrogen, or heat sealed for storage
Specifications
Bulb Draw (Metric)11.6 mL
DescriptionSedi-Pet Transfer Pipette, Narrow Stem, Large Bulb, 15.3mL
Diameter (Metric) Outer3.0 mm
Drops/mL19
GraduatedNo
Packaging TypeBulk
QuantityBulk 250/Pk 10Pk/Cs 2500/Cs
DisposableSingle-use
Length (Metric)15.5 cm
MaterialLDPE
SterilityNon-sterile
TypeNarrow Stem Transfer Pipet
Volume (Metric)15.3 mL
Unit SizeCase of 2500

Citations & References (10)

Citations & References
Abstract
Characterization of the SIM-A9 cell line as a model of activated microglia in the context of neuropathic pain.
Authors:Dave KM, Ali L, Manickam DS
Journal:PLoS One
PubMed ID:32287325
'Resident microglia of the central nervous system are being increasingly recognized as key players in diseases such as neuropathic pain. Biochemical and behavioral studies in neuropathic pain rodent models have documented compelling evidence of the critical role of ATP mediated-P2X4R-brain-derived neurotrophic factor (BDNF) signaling pathway in the initiation and maintenance ... More
Expression of the neonatal Fc-receptor in placental-fetal endothelium and in cells of the placental immune system.
Authors:Kiskova T, Mytsko Y, Schepelmann M, Helmer H, Fuchs R, Miedl H, Wadsack C, Ellinger I
Journal:Placenta
PubMed ID:30955709
'Starting from the second trimester of pregnancy, passive immunity is provided to the human fetus by transplacental transfer of maternal IgG. IgG transfer depends on the neonatal Fc receptor, FcRn. While FcRn localization in the placental syncytiotrophoblast (STB) has been demonstrated unequivocally, FcRn expression in placental-fetal endothelial cells (pFECs), which ... More
Sleep-restriction Inhibits Neurogenesis Through Decreasing the Infiltration of CD169
Authors:Yao H, Zhang Y, Xie B, Shang Y, Yuan S, Zhang J
Journal:Neuroscience
PubMed ID:32081723
'Chronic sleep-restriction (SR) is shown to be correlated with neurodevelopmental disorders. However, the effects of SR during stroke recovery on neurorepair remain unclear. In this study, mice were subjected to 60?min of cerebral ischemia followed by reperfusion. The SR protocol was accomplished by depriving mice of sleep for 20?h/day for ... More
CRISPR Interference-Based Platform for Multimodal Genetic Screens in Human iPSC-Derived Neurons.
Authors:Tian R, Gachechiladze MA, Ludwig CH, Laurie MT, Hong JY, Nathaniel D, Prabhu AV, Fernandopulle MS, Patel R, Abshari M, Ward ME, Kampmann M
Journal:Neuron
PubMed ID:31422865
CRISPR/Cas9-based functional genomics have transformed our ability to elucidate mammalian cell biology. However, most previous CRISPR-based screens were conducted in cancer cell lines rather than healthy, differentiated cells. Here, we describe a CRISPR interference (CRISPRi)-based platform for genetic screens in human neurons derived from induced pluripotent stem cells (iPSCs). We ... More
The WNT/ß-catenin signaling inhibitor XAV939 enhances the elimination of LNCaP and PC-3 prostate cancer cells by prostate cancer patient lymphocytes in vitro.
Authors:Stakheev D, Taborska P, Strizova Z, Podrazil M, Bartunkova J, Smrz D
Journal:Sci Rep
PubMed ID:30886380
Upregulated Wnt/ß-catenin signaling is associated with increased cancer cell resistance and cancer cell-elicited immunosuppression. In non-neoplastic immune cells, upregulated Wnt/ß-catenin is, however, associated with either immunosuppression or immunostimulation. Therefore, it is difficult to predict the therapeutic impact inhibitors of Wnt/ß-catenin signaling will have when combined with cancer immunotherapy. Here, we ... More