Nunc™ Disposable Loops and Needles, needle
Nunc™ Disposable Loops and Needles, needle
Nunc™ Disposable Loops and Needles, needle
Nunc™ Disposable Loops and Needles, needle
Nunc™ Disposable Loops and Needles, needle
Thermo Scientific™
Nunc™ Disposable Loops and Needles, needle
Easily identify these color-coded disposable loops and needles, precision molded from nontoxic (USP Class VI) polystyrene material.
Have Questions?
Catalog number 253988PK
also known as 253988
Price (USD)
250.53
Online Exclusive
278.41
Save 27.88 (10%)
Pack of 1000
Price (USD)
250.53
Online Exclusive
278.41
Save 27.88 (10%)
Pack of 1000
Easily identify color-coded Thermo Scientific™ Nunc™ Disposable Loops and Needles, precision molded from nontoxic (USP Class VI) polystyrene material to provide a smooth loop surface. Thermo Scientific Nunc products are high quality plastics for use in biotechnology, pharmaceutical and research laboratories, as well as in the production of vaccines and diagnostic kits.
Features:
  • Radiation sterilized—no need for flame sterilization
  • Hydrophilic treatment during manufacturing process
  • Liquid adherence properties that prevent accidental discharge of droplets
  • Loops available in 10 and 1μL sizes
  • Meet semi-quantitative standard for handling samples
  • Calibration documents available for any batch upon request
Specifications
ColorYellow
DisposableSingle-use
QuantityCarton of 1000
MaterialPolystyrene
Product TypeInoculating Needle
SterilitySterile
Unit SizePack of 1000