(S)-(-)-2-Chloropropionic acid, 98%
(S)-(-)-2-Chloropropionic acid, 98%
(S)-(-)-2-Chloropropionic acid, 98%
Thermo Scientific Chemicals

(S)-(-)-2-Chloropropionic acid, 98%

CAS: 29617-66-1 | C3H4ClO2 | 107.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 254970010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS29617-66-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4340 to 1.4360 (20°C, 589 nm)
Infrared spectrumConforms
Specific optical rotation-14° to -15° (25°C, 589 nm) (neat)
Appearance (Color)Clear light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only