Mercuric acetate, 99+%
Mercuric acetate, 99+%
Mercuric acetate, 99+%
Thermo Scientific Chemicals

Mercuric acetate, 99+%

CAS: 1600-27-7 | C4H6HgO4 | 318.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
1 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 255651000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1600-27-7
IUPAC Namemercury(2+) diacetate
Molecular FormulaC4H6HgO4
InChI KeyBRMYZIKAHFEUFJ-UHFFFAOYSA-L
SMILES[Hg++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<20 ppm
Residue after ignition=<0.02 % (substances not reducible with formic acid)
Chloride (Cl)=<50 ppm
Nitrate (NO3)=<50 ppm
Sulfate (SO4)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only