3-Methoxypropylamine, 99+%, Thermo Scientific Chemicals
3-Methoxypropylamine, 99+%, Thermo Scientific Chemicals
3-Methoxypropylamine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methoxypropylamine, 99+%, Thermo Scientific Chemicals

CAS: 5332-73-0 | Molecular Formula: C4H11NO | Molecular Weight: 89.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
2575200101 LGlass bottle
Catalog number 257520010
Price (USD)
46.65
Online exclusive
52.20
Save 5.55
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
46.65
Online exclusive
52.20
Save 5.55
Each
Chemical Identifiers
CAS5332-73-0
IUPAC Name3-methoxypropan-1-amine
Molecular FormulaC4H11NO
InChI KeyFAXDZWQIWUSWJH-UHFFFAOYSA-N
SMILESCOCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4165 to 1.4185 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=99.0 %
Appearance (Color)Clear colorless to slightly colored
Infrared spectrumConforms

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only