2-Butoxyacetic acid, 98%
2-Butoxyacetic acid, 98%
2-Butoxyacetic acid, 98%
Thermo Scientific Chemicals

2-Butoxyacetic acid, 98%

CAS: 2516-93-0 | C6H12O3 | 132.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
2577600101 gGlass bottle
Catalog number 257760010
Price (TWD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2516-93-0
IUPAC Namesodium 2-butoxyacetate
Molecular FormulaC6H11NaO3
InChI KeyVFOVBGNGALJINB-UHFFFAOYSA-M
SMILES[Na+].CCCCOCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4250 to 1.4270 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only