Buffer solution pH 7, Phosphate buffer - FAQs
Buffer solution pH 7, Phosphate buffer - FAQs
View additional product information for Buffer solution pH 7, Phosphate buffer - FAQs (258595000, 258590010, 258591000, 258590025)
2 product FAQs found
What is the concentration of the Buffer solution pH 7, Phosphate buffer?
The Buffer solution pH 7, Phosphate buffer has a concentration of 0.1 M.
What is the composition of the Buffer solution pH 7, Phosphate buffer?
The composition of the Buffer solution pH 7, Phosphate buffer is 3.54 g potassium dihydrogen phosphate and 14.7 g di-sodium hydrogen phosphate per liter.