2-Phenylbenzimidazole-5-sulfonic acid, 98%
2-Phenylbenzimidazole-5-sulfonic acid, 98%
2-Phenylbenzimidazole-5-sulfonic acid, 98%
Thermo Scientific Chemicals

2-Phenylbenzimidazole-5-sulfonic acid, 98%

CAS: 27503-81-7 | C13H10N2O3S | 274.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 259700250
Price (USD)
37.65
Special offer
Online exclusive
Ends: 28-Dec-2026
44.60
Save 6.95
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
37.65
Special offer
Online exclusive
Ends: 28-Dec-2026
44.60
Save 6.95
Each
Chemical Identifiers
CAS27503-81-7
IUPAC Name2-phenyl-1H-1,3-benzodiazole-6-sulfonic acid
Molecular FormulaC13H10N2O3S
InChI KeyUVCJGUGAGLDPAA-UHFFFAOYSA-N
SMILESOS(=O)(=O)C1=CC=C2N=C(NC2=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only