Sodium hydroxide, extra pure, 50 wt% solution in water
Sodium hydroxide, extra pure, 50 wt% solution in water
Sodium hydroxide, extra pure, 50 wt% solution in water
Thermo Scientific Chemicals

Sodium hydroxide, extra pure, 50 wt% solution in water

CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
2.5 L
5 L
25 L
Packaging:
View all
Plastic Bottle
Catalog number 259860025
Price (USD)
126.65
Online exclusive
140.00
Save 13.35
Each
Quantity:
2.5 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
126.65
Online exclusive
140.00
Save 13.35
Each
Chemical Identifiers
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HCl49.0 to 51.0 %
Density(20°C) 1.510 to 1.540 kg/l
Sodium chloride (NaCl)=<200 ppm (NaCl)
Heavy metals=<10 ppm
Appearance (Form)Viscous solution
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only