L(+)-Selenomethionine, 99+%, ee 99+%
L(+)-Selenomethionine, 99+%, ee 99+%
L(+)-Selenomethionine, 99+%, ee 99+%
Thermo Scientific Chemicals

L(+)-Selenomethionine, 99+%, ee 99+%

CAS: 3211-76-5 | C5H11NO2Se | 196.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
2.5 g
Packaging:
Glass bottle
Catalog number 259960010
Price (USD)
292.65
Special offer
Online exclusive
Ends: 28-Dec-2026
344.00
Save 51.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
292.65
Special offer
Online exclusive
Ends: 28-Dec-2026
344.00
Save 51.35
Each
Chemical Identifiers
CAS3211-76-5
IUPAC Name(2S)-2-amino-4-(methylselanyl)butanoic acid
Molecular FormulaC5H11NO2Se
InChI KeyRJFAYQIBOAGBLC-BYPYZUCNSA-N
SMILESC[Se]CC[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Enantiomeric excess>=99.0 % (HPLC-CHIR)
Titration with HClO499.0 to 100.5 %
Appearance (Form)Glistening powder
Appearance (Color)White to off-white
HPLC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only