Fluoraldehyde™ o-Phthaldialdehyde Reagent Solution (OPA) - FAQs
View additional product information for Fluoraldehyde™ o-Phthaldialdehyde Reagent Solution (OPA) - FAQs (26025)
1 product FAQs found
What is the shelf life for Fluoraldehyde o-Phthaldialdehyde Reagent Solution (OPA) (Cat. No. 26025)?
The lot-specific expiration date for Fluoraldehyde o-Phthaldialdehyde Reagent Solution (OPA) (Cat. No. 26025) can be found on the Certificate of Analysis.