Pierce™ Control Agarose Resin
Thermo Scientific™

Pierce™ Control Agarose Resin

Thermo Scientific Control Agarose Resin is underivatized beaded agarose support for pre-clearing protein immunoprecipitation samples to reduce background signal andRead more
Have Questions?
Catalog NumberQuantity
2615010 mL
Catalog number 26150
Price (USD)
174.65
Online exclusive
199.00
Save 24.35
Each
Quantity:
10 mL
Request bulk or custom format
Price (USD)
174.65
Online exclusive
199.00
Save 24.35
Each
Thermo Scientific Control Agarose Resin is underivatized beaded agarose support for pre-clearing protein immunoprecipitation samples to reduce background signal and nonspecific binding in IP and Co-IP experiments.

The unmodified and non-activated Control Agarose Resin is the same support that is included in our immunoprecipitation (IP) kits and co-immunoprecipitation (co-IP) kits. The resin is effective for depleting proteins and other sample components that bind to agarose nonspecifically, which reduces background signal in IP reactions. When used as a negative control, these beads enable specific and nonspecific binding interactions to be distinguished.

Features of Control Agarose Resin:

• Matched control resin—the crosslinked 4% beaded agarose (CL4B) is the same base support used in our various IP and co-IP kits
• Ideal package size—unlike other suppliers who offer only 1 liter bottles of similar agarose resin, our 10 mL package size is more appropriate for experimental needs
• Convenient format—the resin is supplied as a 50% slurry in 20% ethanol (pipetting 1 mL of slurry delivers 0.5 mL of settled resin); just wash in buffer and the beads are ready to use
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column TypeAgarose Resin
FormatBottle
Quantity10 mL
FormLiquid Suspension
Product LinePierce
TypeResin
Unit SizeEach
Contents & Storage
Store at 2–8°C.