Proteinase K Solution, ChIP grade - FAQs
Proteinase K Solution, ChIP grade - FAQs
View additional product information for Proteinase K Solution, ChIP grade - FAQs (26160)
1 product FAQs found
How do you recommend storing proteinase K?
We recommend storing it at 4°C.