Nunc™ Microplate Lids
Nunc™ Microplate Lids
Nunc™ Microplate Lids
Nunc™ Microplate Lids
Thermo Scientific™

Nunc™ Microplate Lids

Protect samples from contamination and evaporation during assay processing, incubation or storage with these microplate lids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPlate CompatibilitySterilityCondensation RingsNo. per Pack
26333996-well PlateNon-sterileYes5
250002All MicroplatesSterileNo20
250003All MicroplatesNon-sterileNo20
250005All MicroplatesNon-sterileNo20
253623All MicroplatesNon-sterileYes25
253624All MicroplatesSterileYes25
26412296-well PlateSterileYes1
264611384-well PlateSterileNo1
264612384-well PlateNon-sterileNo20
264616384-well PlateSterileNo20
26462396-well PlateSterileYes20
Catalog number 263339
Price (USD)
264.76
Online exclusive
308.53
Save 43.77
Case of 100
Plate Compatibility:
96-well Plate
Sterility:
Non-sterile
Condensation Rings:
Yes
No. per Pack:
5
Price (USD)
264.76
Online exclusive
308.53
Save 43.77
Case of 100
Protect samples from contamination and evaporation during assay processing, incubation or storage with Thermo ScientificTM NuncTM Microplate Lids.
  • Lids are for use with any standard plates
  • 96 Well Lids have individual condensation rings
  • Lids have notches for quick orientation
  • Universal Lids can be used on any standard height plate on the market
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Condensation RingsYes
Cut-off CornersLong Side
Dimensions (L x W)5 x 3.35 in. (127 x 85 mm)
Dimensions (L x W x H)5 x 3.35 x 0.3 in. (127 x 85 x 7.9 mm)
Evaporation BarrierYes
Height (English)0.3 in.
Height (Metric)7.9 mm
Length (English)5 in.
Length (Metric)127 mm
No. per Case100
No. per Pack5
Product TypePlate Lid
Temperature Range (Metric)-20°C to +80°C
Well ShapeAll well shapes
Width (English)3.35 in.
Width (Metric)85 mm
AdhesiveNone
ColorClear
MaterialPolystyrene
No. of Wells96
Plate Compatibility96-well Plate
Product LineNunc
SterilityNon-sterile
Unit SizeCase of 100