L(-)-Camphorsulfonic acid, 98%
L(-)-Camphorsulfonic acid, 98%
L(-)-Camphorsulfonic acid, 98%
Thermo Scientific Chemicals

L(-)-Camphorsulfonic acid, 98%

CAS: 35963-20-3 | C10H16O4S | 232.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 263645000
Price (USD)
391.65
Online Exclusive
435.00
Save 43.35 (10%)
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
391.65
Online Exclusive
435.00
Save 43.35 (10%)
Each
Chemical Identifiers
CAS35963-20-3
IUPAC Name[(1R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonic acid
Molecular FormulaC10H16O4S
InChI KeyMIOPJNTWMNEORI-MHPPCMCBSA-N
SMILESCC1(C)C2CC[C@]1(CS(O)(=O)=O)C(=O)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=97.5 %
Appearance (Form)Crystalline powder
Appearance (Color)White to light beige
Infrared spectrumConforms
Specific optical rotation-21° to -23° (20°C, 589 nm) (c=20, H2O)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only