Potassium phosphate, tribasic monohydrate, 96%, extra pure
Potassium phosphate, tribasic monohydrate, 96%, extra pure
Potassium phosphate, tribasic monohydrate, 96%, extra pure
Thermo Scientific Chemicals

Potassium phosphate, tribasic monohydrate, 96%, extra pure

CAS: 27176-10-9 | K3O4P·H2O | 230.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 263795000
Price (USD)
54.65
Online exclusive
60.60
Save 5.95
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
54.65
Online exclusive
60.60
Save 5.95
Each
Chemical Identifiers
CAS27176-10-9
IUPAC Nametripotassium hydrate phosphate
Molecular FormulaH2K3O5P
InChI KeyRMNIZOOYFMNEJJ-UHFFFAOYSA-K
SMILESO.[K+].[K+].[K+].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals or granules
Loss on ignition=<13 % (800°C)
Appearance (Color)White
Titration excess HCl>=95 %
Sulfate (SO4)=<200 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only