(S)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, 99%
(S)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, 99%
(S)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, 99%
Thermo Scientific Chemicals

(S)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, 99%

CAS: 20445-33-4 | C10H8ClF3O2 | 252.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number 264112500
Price (USD)
153.65
Online Exclusive
170.00
Save 16.35 (10%)
Each
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
153.65
Online Exclusive
170.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS20445-33-4
IUPAC Name(2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride
Molecular FormulaC10H8ClF3O2
InChI KeyPAORVUMOXXAMPL-SECBINFHSA-N
SMILESCO[C@@](C(Cl)=O)(C1=CC=CC=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4680 to 1.4700 (20°C, 589 nm)
GC>=98.5 %
Specific optical rotation+130° to +134° (20°C, 589 nm) (c=6, CHCl3)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chiral derivatization reagent
RUO – Research Use Only