Ytterbium(III) oxide, 99.99%, (trace metal basis)
Ytterbium(III) oxide, 99.99%, (trace metal basis)
Ytterbium(III) oxide, 99.99%, (trace metal basis)
Thermo Scientific Chemicals

Ytterbium(III) oxide, 99.99%, (trace metal basis)

CAS: 1314-37-0 | O3Yb2 | 394.09 g/mol
製品番号(カタログ番号) 264300100
価格(JPY)
-
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1314-37-0
IUPAC Namediytterbium(3+) trioxidandiide
Molecular FormulaO3Yb2
InChI KeyUZLYXNNZYFBAQO-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Yb+3].[Yb+3]
さらに表示
Appearance (Form)Powder
Total trace metal impurities=<100 ppm
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only