Nunc™ 384-Well Polypropylene Sample Processing & Storage Microplates
Nunc™ 384-Well Polypropylene Sample Processing & Storage Microplates
Nunc™ 384-Well Polypropylene Sample Processing & Storage Microplates
Nunc™ 384-Well Polypropylene Sample Processing & Storage Microplates
Thermo Scientific™
Nunc™ 384-Well Polypropylene Sample Processing & Storage Microplates
Green features
Save on costs when storing compounds for small volume applications. Plates feature lower binding capacity so proteins or DNA will not bind, allowing for complete sample recovery.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSterilityWell ShapeFit for Purpose
264579BlueNon-sterileSquare, Round Bottom (U-well)Certified Quality
264573NaturalNon-sterileSquare, Round Bottom (U-well)Certified Quality
264574NaturalSterileSquare, Round Bottom (U-well)Certified Quality
264575WhiteNon-sterileSquare, Conical Bottom (V-well)Certified Quality
264576BlackNon-sterileSquare, Round Bottom (U-well)Certified Quality
4309NaturalSterileSquare, Conical Bottom (V-well)Certified Quality
4308BlackNon-sterileSquare, Conical Bottom (V-well)Certified Quality
4312NaturalNon-sterileSquare-RoundedCertified Quality
Catalog number 264579
Price (USD)
992.97
Online exclusive
1,158.12
Save 165.15
Case of 120
Color:
Blue
Sterility:
Non-sterile
Well Shape:
Square, Round Bottom (U-well)
Fit for Purpose:
Certified Quality
Price (USD)
992.97
Online exclusive
1,158.12
Save 165.15
Case of 120
Minimize costs when storing compounds for small volume applications. Thermo Scientific™ 384-Well Conical Bottom Polypropylene Plates feature a lower binding capacity so proteins or DNA will not bind, allowing for complete sample recovery. Withstands temperatures from -80° to +121°C. Offered in a variety of colors for quick identification of your storage plates.
  • Conical wells optimize sample recovery
  • Square rounded wells minimize wicking (capillary action)
  • Chemical resistant to DMSO
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
AutoclavableAutoclavable
Binding TypeLow binding PP
Certifications/ComplianceCertified RNase and DNase Free. ANSI Compliant.
Dnase FreeYes
Description384-well Polypropylene Sample Processing & Storage Plates
For Use With (Application)Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
For Use With (Equipment)ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
FormatStandard
LidWithout Lid
No. per Case120
No. per Pack20
Packaging120 Cases
Plate SizeANSI/ SLAS Standard
Product Type384-well plate
Quantity120/Cs.
RNase FreeYes
Regulatory StatusRUO
Volume (Metric) Well120 μL
Volume (Metric) Working100 μL
Array16 x 24
BarcodeNo Barcode
ColorBlue
Fit for PurposeCertified Quality
MaterialPolypropylene
No. of Wells384
Product LineNunc
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeSquare, Round Bottom (U-well)
Unit SizeCase of 120