Nunc™ 384 Well ShallowWell Standard Height Black and White Plates
Thermo Scientific™
Nunc™ 384 Well ShallowWell Standard Height Black and White Plates
Save costs in samples and reagents by reducing the total well volume to 25μL using Nunc 384-well shallow well standard height black and white polystyrene plates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSurface Treatment
264706WhiteNon-treated
264705BlackNon-treated
Catalog number 264706
Price (USD)
1,198.00
Case of 100
Color:
White
Surface Treatment:
Non-treated
Price (USD)
1,198.00
Case of 100
Accommodate small sample volumes by using Thermo Scientific™ Nunc™ 384-Well ShallowWell Standard Height Black and White Plates with total well volume of 25μL, ideal for high throughput screenings. The standard plate height allows for easy adaptation to most automation systems.
Features:
  • Black color minimizes back-scattered light and background noise
  • ANSI™ (SBS) standard footprint
  • Customized bar coding available
Warranty: 90 days
WARNING: Cancer – www.P65Warnings.ca.gov
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
FormatStandard
LidWithout Lid
No. per Case100
No. per Pack25
Packaging100 Cases, 25 Pk.
Product TypeMicroplate
ProfileStandard Height
Volume (Metric) Well25 μL
Volume (Metric) Working2 to 20 μL
Well DesignShallow Well
Array16 x 24
ColorWhite
MaterialPolystyrene
No. of Wells384
Product LineNunc
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeFlat Bottom (F-well)
Unit SizeCase of 100
Frequently asked questions (FAQs)
Can I use Nunc White 384-Well Microplates for fluorescent assays?
Nunc White 384-Well Microplates can be used for fluorescent applications, but background fluorescence will be much higher compared to Nunc Black 384-Well Microplates.
We recommend the white microplates for scintillation and luminescent detection systems, in which the reflective properties of the microplate enhance the signal without producing any endogenous background signal.
Do you offer Nunc Microplate Lids for the Nunc Black or White 384-Well Microplates?
We offer Nunc Microplate Lids: Cat. No. 448143, that fit both the 384-well microplates and the 1,536-well microplates.
Why do you offer Nunc 384-Well Black or White Microplates when the cells cannot be observed using an inverted light microscope?
We offer Nunc Black or White 384-Well microplates that are intended for use in conjunction with 384-well microplate luminometers and fluorometers that have excitation and detection systems located above the plate. Due to the opaque quality of these microplates, all detection systems must rely on either luminescent or fluorescent reagents, such as viability stains that report cell activity and membrane integrity.