Tris-phosphate, 98%
Tris-phosphate, 98%
Tris-phosphate, 98%
Thermo Scientific Chemicals

Tris-phosphate, 98%

CAS: 6992-39-8 | C4H14NO7P | 219.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
264952500250 gGlass bottle
Catalog number 264952500
Price (USD)
253.65
Special offer
Online exclusive
Ends: 28-Dec-2026
298.00
Save 44.35
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
253.65
Special offer
Online exclusive
Ends: 28-Dec-2026
298.00
Save 44.35
Each
Chemical Identifiers
CAS6992-39-8
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol; phosphoric acid
Molecular FormulaC4H14NO7P
InChI KeyJLEXUIVKURIPFI-UHFFFAOYSA-N
SMILESOP(O)(O)=O.NC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point168°C to 172°C
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only