Nalgene™ Closed-Dome Polypropylene Tanks
Thermo Scientific™

Nalgene™ Closed-Dome Polypropylene Tanks

Store, mix and dispense reagents in Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
2650-0100378.5 L
2650-002075 L
2650-0030115 L
2650-0055210 L
Catalog number 2650-0100
Price (USD)
2,705.00
Each
Capacity (Metric):
378.5 L
Price (USD)
2,705.00
Each

Store, mix and dispense reagents in Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks, designed for use as closed containment systems. Offered in sizes ranging from 75 to 380L , tanks feature a 150mm neck opening fitted with a gasketed closure for secure sealing.

Features:

  • Tanks may be fitted with spigots
  • Autoclavable tanks and closures feature polypropylene construction

Compatible with:
Use with Closed-Dome Tank Closure w/ Mixer Support Assembly (Mfr. No. 2651-0200), BioTech Overhead Mixer (Mfr. No. 2653-0010 and 2653-0020) and Lower Assemblies for BioTech Mixers (Mfr. No. 2654-0030, 2654-0055, 2654-0075 & 2654-0100).

Warranty:
90 days

Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Specifications
Capacity (English)100 gal.
Certifications/ComplianceComply with FDA Reg. 177.1520 and USP Class VI
Diameter (English) Outer28.5 in.
Height (English)52 in.
Height (Metric)132.1 cm
No. per Case1
No. per Pack1
Capacity (Metric)378.5 L
Diameter (Metric) Outer72 cm
MaterialPolypropylene
TypeClosed-dome Tank
Unit SizeEach