Product Image
Thermo Scientific™
Nunc™ 384-Well, Non-Treated, Flat-Bottom Microplate
Thermo Scientific™ Nunc 384-Well Clear Polystyrene Plates with Cell Culture Treated and Nontreated Surfaces are optimized for robotics, plate readers and liquid handling systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of WellsNo. per PackNo. per Case
26520238425 Microplates100 Microplates
24275738410 Microplates30 Microplates
242765961 Microplates30 Microplates
Catalog number 265202
Price (USD)
824.65
Online exclusive
850.00
Save 25.35
Case of 100
No. of Wells:
384
No. per Pack:
25 Microplates
No. per Case:
100 Microplates
Price (USD)
824.65
Online exclusive
850.00
Save 25.35
Case of 100
Thermo Scientific™ Nunc 384-Well Clear Polystyrene Plates with Cell Culture Treated and Nontreated Surfaces are optimized for robotics, plate readers and liquid handling systems. 384-well microplates allow sample and reagent volume reduction providing cost savings and allowing more samples to be screened. Our plates are high quality for consistent and reliable results.
Nunc 384-well polystyrene plates have rounded square-well geometry, eliminating wicking.
  • Clear Polystyrene has excellent optical properties making it ideal for optical measurements
  • Untreated Polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
  • Untreated surfaces are ideal for adding coatings in your lab
  • Cell Culture Treated: The Nunclon Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Customized bar coding is available
  • Flexible working volumes; 10-100 μL
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Culture Area0.084 cm2/well
DescriptionNon-Treated; w/ lid; Sterile; 25/100
FormatSBS Standard
LidWith Lid
No. per Case100 Microplates
No. per Pack25 Microplates
Packaging25/Pk., 100/Cs.
Product TypeMicroplate
Quantity25 Pk.
Volume (Metric) Well120 μL
Volume (Metric) Working10 to 100 μL
Array16 x 24
BarcodeCustom Barcoding Available
ColorClear
MaterialPolystyrene
No. of Wells384
Product LineNunc
SterilitySterile
Surface TreatmentNon-treated
Well ShapeFlat Bottom (F-well)
Unit SizeCase of 100
Frequently asked questions (FAQs)
Should I use an opaque black/white immuno plate, or an "optical-bottom" design?
Opaque plates can only be used on “top reader” instruments, which detect (or, in fluorescence mode, excite and detect) from above the plate. Readers that detect from underneath the plate (or imaging systems) use optical-bottom plates, which have a black or white upper well structure and a transparent (polystyrene film or optical coverglass) bottom.
Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.