96 Black Plate, Non-Treated Coverglass Surface, No Lid, Non-Sterile, Pack of 5
Thermo Scientific™
96 Black Plate, Non-Treated Coverglass Surface, No Lid, Non-Sterile, Pack of 5
Color:Black, Clear Bottom; Sterility; Non-sterile; Lid, Without Lid; Type: Optical Coverglass Base
Have Questions?
카탈로그 번호수량
265300Case of 30
카탈로그 번호 265300
제품 가격(KRW)
2,614,000
온라인 행사
Ends: 31-Dec-2026
2,904,000
할인액 290,000 (10%)
Case of 30
수량:
Case of 30
제품 가격(KRW)
2,614,000
온라인 행사
Ends: 31-Dec-2026
2,904,000
할인액 290,000 (10%)
Case of 30
Achieve optimal fluorescence and luminescence in cell culture and microscopic applications with the Thermo Scientific™ Nunc™ MicroWell™ 96-Well Optical-Bottom Plates with Coverglass Base, featuring a polystyrene black or white upper structure with a polystyrene film at the bottom of the plate.
Features:
  • Background, autoflorescence and crosstalk are improved
  • Black microplates for florescence measurements, with minimum back-scattered light and background fluorescence
  • White microplates for luminescence measurements, with maximum reflection and minimal autoluminescence
  • Cell culture surface is ideal for cell-based assays
  • CC2 surface modification closely mimics Poly-D-Lysine and is superior for attachment and growth of fastidious cells
  • Customized bar coding available
사양
인증/적합성All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
뚜껑Without Lid
케이스당 수30
팩당 수5
포장5/Pk., 30/Cs.
제품 유형Optical Coverglass Base
수량Case of 30
어레이8 x 12
바코드Custom Barcoding Available
색상Black
물질Polystyrene
웰 수96
제품라인Nunc
멸균Non-sterile
표면 처리Non-treated
웰 형상Flat Bottom (F-well)
Unit SizeCase of 30
자주 묻는 질문(FAQ)
Should I use an opaque black/white immuno plate, or an "optical-bottom" design?
Opaque plates can only be used on “top reader” instruments, which detect (or, in fluorescence mode, excite and detect) from above the plate. Readers that detect from underneath the plate (or imaging systems) use optical-bottom plates, which have a black or white upper well structure and a transparent (polystyrene film or optical coverglass) bottom.
Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.