(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 99+%
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 99+%
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 99+%
Thermo Scientific Chemicals

(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 99+%

CAS: 76189-56-5 | C44H32P2 | 622.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 265540050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS76189-56-5
IUPAC Name[2'-(diphenylphosphanyl)-[1,1'-binaphthalen]-2-yl]diphenylphosphane
Molecular FormulaC44H32P2
InChI KeyMUALRAIOVNYAIW-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1C1=C(C=CC2=CC=CC=C12)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White to light yellow
Infrared spectrumConforms
Melting point238°C to 242°C
HPLC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only