I used one of your PageRuler unstained protein ladders and did not get good separation of the bands. What could have happened?
Here are possible causes and solutions:
- Ladder was boiled: Discard boiled aliquot.
- Too much volume of ladder used: Add less volume or dilute the ladder in protein loading buffer prior to use.
- DTT oxidation in storage buffer: Add freshly prepared DTT solution to a final concentration of 100 mM.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
- Ladder was boiled: Discard boiled aliquot.
- Too much volume of ladder used: Add less volume or dilute the ladder in protein loading buffer prior to use.
- DTT oxidation in storage buffer: Add freshly prepared DTT solution to a final concentration of 100 mM.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.