Nunc™ Rectangular Dishes
Nunc™ Rectangular Dishes
Nunc™ Rectangular Dishes
Thermo Scientific™

Nunc™ Rectangular Dishes

Perform molecular biology applications with non-treated Thermo Scientific™ Nunc™ Rectangular Dishes, which have ANSI™ standard dimensions and are optimized for robotics.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Wells
2670601
2670614
2670628
Catalog number 267060
Price (USD)
577.65
Online exclusive
614.00
Save 36.35
Case of 100
No. of Wells:
1
Price (USD)
577.65
Online exclusive
614.00
Save 36.35
Case of 100
Perform molecular biology applications with non-treated Thermo Scientific™ Nunc™ Rectangular Dishes, which have ANSI™ standard dimensions and are optimized for robotics.

Rectangular Dishes 1, 4 and 8-Well

  • 1-well dish is ideal for HTS Petri dish applications
  • 4- and 8-well dishes are ideal for assay compartmentalization, probe multiple gene sub-set
  • 4-well dish holds four glass slides for storage

1-Well Format

  • Convenient plate format for cell culture
  • HTS Petri dish applications

4- and 8-Well Formats

  • Assay compartmentalization
  • Probe multiple gene subsets
  • Wash Western blot strips
  • 4-well dish holds four glass slides
  • Useful in all areas of cell culture, including scale-up and cloning

Ordering Information:

Customized bar coding is available

WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
FormatSBS Standard
IncludesLid
Length (Metric)100 mm
No. per Case100/Cs.
No. per Pack10 Pk.
Product TypeCell Culture Dish
Quantity10 Pk.
Total Volume (Metric) Well90 mL
Width (Metric)86 mm
Array1 x 1
BarcodeNo Barcode
ColorClear
MaterialPolystyrene
No. of Wells1
SterilitySterile
Surface TreatmentUntreated
Well ShapeRectangular
Unit SizeCase of 100